Schwefelsäure ≥95%, Finyte® für Mikroelektronik, J.T.Baker®

Lieferant: Avantor

5802-15EA 332 EUR
5802-15
Schwefelsäure ≥95%, Finyte® für Mikroelektronik, J.T.Baker®
Schwefelsäure
Formel: H₂SO₄
Molecular Weight: 98,08 g/mol
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
UN: 1830
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use 95.0 - 97.0 %
Recommended Storage Conditions: 15° - 100°F = 10
Assay (H₂SO₄) 95.0 - 97.0 %
Color (APHA) ≤10
Chloride (Cl) ≤0.1 ppm
Nitrate (NO₃) ≤0.1 ppm
Phosphate (PO₄) ≤0.20 ppm
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Arsenic and Antimony (as As) ≤2.0 ppb
Trace Impurities - Barium (Ba) ≤5.0 ppb
Trace Impurities - Beryllium (Be) ≤1.0 ppb
Trace Impurities - Bismuth (Bi) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤5.0 ppb
Trace Impurities - Calcium (Ca) ≤10.0 ppb
Trace Impurities - Chromium (Cr) ≤5.0 ppb
Trace Impurities - Cobalt (Co) ≤5.0 ppb
Trace Impurities - Copper (Cu) ≤5.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤5.0 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤5.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤5.0 ppb
Trace Impurities - Niobium (Nb) ≤5.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silicon (Si) ≤10.0 ppb
Trace Impurities - Silver (Ag) ≤5.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤5.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤5.0 ppb
Trace Impurities - Vanadium (V) ≤5.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and greater ≤25 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR