TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid), Millipore®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: TCEP HCl

580560-1
580560-1EA 137 EUR
580560-1
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid), Millipore®
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
Formel: C₉H₁₆ClO₆P
Molecular Weight: 286.65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00145469
CAS-Nummer: 51805-45-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR