TCEP, neutral, Calbiochem®
Lieferant: Merck Millipore Calbiochem
580561-1
580561-1EA 186 EUR
580561-1
TCEP, neutral, Calbiochem®
Protein-Aufbereitungs Proteinreinigungs-Reagenzien
A ready-to-use stable, neutral form of the disulfide-reducing agent TCEP, hydrochloride.