4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol
581500-500
581500-500EA
127
EUR
581500-500
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL), Millipore®
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL)
Formel:
C₉H₁₈NO₂ Molecular Weight: 172,25 g/mol |
MDL:
MFCD00006478 CAS-Nummer: 2226-96-2 EINECS: 218-760-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...