ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Lieferant: Merck
58221-500ML-F
58221-500ML-FEA 133 EUR
58221-500ML-F
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Puffer Ionenstärkepuffer
Solution used to establish a constant ionic strength in a sample solution containing e.g. calcium for det. with ISE.
  • Prepared with high purity salts