ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Lieferant: Merck
58221-500ML-F
58221-500ML-FEA
133
EUR
58221-500ML-F
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Puffer
Ionenstärkepuffer
Solution used to establish a constant ionic strength in a sample solution containing e.g. calcium for det. with ISE.
- Prepared with high purity salts