Wasserstoffperoxid 30%, Finyte® für Mikroelektronik, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Veränderungsmanagement-Kategorie= R
5846-03EA
0
EUR
5846-03
Wasserstoffperoxid 30%, Finyte® für Mikroelektronik, J.T.Baker®
Wasserstoffperoxid
Formel:
H₂O₂ Siedepunkt: ∼107 °C (1013 hPa) Schmelzpunkt: < 0 °C Dichte: 1,11 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011333 CAS-Nummer: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1,II |
Spezifikation Testergebnisse
For Microelectronic Use | 30.0 - 32.0 % |
Assay (H₂O₂) | 30.0 - 32.0 % |
Color (APHA) | ≤ 6 |
Free Acid (µeq/g) | ≤ 0.6 |
Residue after Evaporation | ≤ 5 ppm |
Chloride (Cl) | ≤ 0.06 ppm |
Nitrate (NO₃) | ≤ 0.4 ppm |
Phosphate (PO₄) | ≤ 0.20 ppm |
Sulfate (SO₄) | ≤ 0.2 ppm |
Trace Impurities - Aluminum (Al) | ≤ 10.0 ppb |
Arsenic and Antimony (as As) | ≤ 5.0 ppb |
Trace Impurities - Barium (Ba) | ≤ 10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤ 10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤ 10.0 ppb |
Trace Impurities - Boron (B) | ≤ 10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤ 10.0 ppb |
Trace Impurities - Calcium (Ca) | ≤ 10.0 ppb |
Trace Impurities - Chromium (Cr) | ≤ 10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤ 10.0 ppb |
Trace Impurities - Copper (Cu) | ≤ 10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤ 10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤ 10.0 ppb |
Trace Impurities - Gold (Au) | ≤ 10.0 ppb |
Trace Impurities - Iron (Fe) | ≤ 10.0 ppb |
Trace Impurities - Lead (Pb) | ≤ 10.0 ppb |
Trace Impurities - Lithium (Li) | ≤ 10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤ 10.0 ppb |
Trace Impurities - Manganese (Mn) | ≤ 10.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤ 10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤ 10.0 ppb |
Trace Impurities - Niobium (Nb) | ≤ 10.0 ppb |
Trace Impurities - Potassium (K) | ≤ 10.0 ppb |
Trace Impurities - Silicon (Si) | ≤ 10.0 ppb |
Trace Impurities - Silver (Ag) | ≤ 10.0 ppb |
Trace Impurities - Sodium (Na) | ≤ 10.0 ppb |
Trace Impurities - Strontium (Sr) | ≤ 10.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤ 10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤ 10.0 ppb |
Trace Impurities - Tin (Sn) | ≤ 10.0 ppb |
Trace Impurities - Titanium (Ti) | ≤ 10.0 ppb |
Trace Impurities - Vanadium (V) | ≤ 10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤ 10.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤ 10.0 ppb |
Particle Count - 0.2 µm and greater | ≤ 1175 par/ml |
Particle Count - 0.5 µm and greater | ≤ 25 par/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...