Supelco® Ascentis® Express Phenyl-Hexyl HPLC Columns
Lieferant: Merck
584621-U 584622-U 584609-U 584610-U 584611-U 584612-U 584613-U 584614-U 584615-U 584616-U 584617-U 584618-U 584619-U 584620-U 584623-U
584621-UEA 571 EUR
584621-U 584622-U 584609-U 584610-U 584611-U 584612-U 584613-U 584614-U 584615-U 584616-U 584617-U 584618-U 584619-U 584620-U 584623-U
Supelco® Ascentis® Express Phenyl-Hexyl HPLC Columns
Chromatographiesäulen
Ascentis® Express 160 Å Phenyl-Hexyl high-speed, high-performance liquid chromatography columns are based on a 160 Å Fused-Core® particle design.
The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0,5 μm thick porous shell and the small overall particle size of 2,7 μm.
The endcapped, phenyl-hexyl bonded phase provides a stable, reversed phase packing with a pore structure and pore size that is optimised for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favoured for proteomic applications.
Lieferumfang: Supplied with column test certificate inside the box.