Capillary cutter, CleanCut
Lieferant: S.C.A.T.
900103
590-0850EA 34.9 EUR
590-0850
Capillary cutter, CleanCut
Schneidewerkzeuge
This cutter is made of PP, blue, with stainless steel blade.
  • Separates capillary tubes safely and reliably
  • With useful openings for measuring the capillary diameter
  • Prevents injuries and ensures a clean cut
The universal tube cutter for HPLC capillaries - the tube guide enables a comfortable, straight cut. With spare blade for exchange.