VWR®, Abluftfilter
Lieferant: VWR Collection
590-1511EA 146 EUR
590-1511
VWR®, Abluftfilter
Filter Belüftungsfilter
These exhaust filters adsorb 99% of all volatile substances, vapours and gases, even organic solvents. They can be used on any HPLC system with multiple capillaries and connectors.
It is recommended that you exchange exhaust filter S every three months and the exhaust filter M every six months.