Calcimycin (A23187) freie Säure ≥98% (durch TLC) Ionophor

Lieferant: Biotium
Achtung

Synonyme: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

59001
59001.EA 98.7 EUR
59001.
Calcimycin (A23187) freie Säure ≥98% (durch TLC) Ionophor
Calcimycin (A23187)

A-23187, also known as Calcimycin or Calcium Ionophore III, is a calcium ionophore that rapidly equilibrates intracellular and extracellular calcium concentrations. It is commonly used for in situ calibrations of fluorescent calcium indicators. The ionophore allows Mn2 to enter the cells and quench intracellular fluorescence of calcium indicators.


  • White solid soluble in DMSO
  • Store at 4°C and protect from light


Also see 4-bromo A-23187 (59006). Because 4-bromo A-23187 is nonfluorescent, it is preferred over the blue fluorescent A-23187 for calibrations of UV-excited Ca2 indicators such as Indo-1 and Fura-2 in order to keep background fluorescence to a minimum.

Formel: C₂₉H₃₇N₃O₆
Molecular Weight: 523,63 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00151202
CAS-Nummer: 52665-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR