N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
Lieferant: Biotium
|
Achtung
|
Synonyme:
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimid Hydrochlorid, EDAC HCl, WSC HCl, EDAC Hydrochlorid, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Monohydrochlorid, N'-(Ethylkohlenstoffimidoyl)-N,N-dimethylpropan-1,3-diaminmonohydrochlorid, EDC Hydrochlorid, EDC HCl
59002
59002.EA
145
EUR
59002.
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
Formel:
C₈H₁₇N₃·HCl Molecular Weight: 191.7 g/mol Schmelzpunkt: 112…113 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012503 CAS-Nummer: 25952-53-8 EINECS: 247-361-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...