N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylendiamin ≥95% (durch TLC) Ionen-Chelator
Lieferant: Biotium
TPEN (Tetrakis-(2-pyridylmethyl)ethylenediamine) is an excellent heavy metal chelator while it does not affect Ca2 , Mg2 , Na , or K concentrations. Thus, TPEN is a useful tool to identify effects of heavy metals (Zn2 , Fe2 /Fe3 , Cu2 , and Mn2 , etc.) on fluorescent indicators of Ca2 , Mg2 , Na , or K .
- Cell permeability: Membrane permeant
- Off-white solid soluble in ethanol, DMSO or DMF
- Store at 4°C
Formel:
C₂₆H₂₈N₆ Lagertemperatur: Kühlschrank |
MDL:
MFCD00036918 CAS-Nummer: 16858-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...