4-Bromo-A-23187 freie Säure ≥98% (durch TLC, NMR) Ionophor, zur Kalziumbestimmung

Lieferant: Biotium

Synonyme: Antibiotic A-23187, 4-bromo

59006
59006.EA 303 EUR
59006.
4-Bromo-A-23187 freie Säure ≥98% (durch TLC, NMR) Ionophor, zur Kalziumbestimmung
4-Bromo-A-23187

Similar to A-23187, 4-Bromo A-23187 is used for in situ calibrations of fluorescent Ca2 indicators to equilibrate intracellular and extracellular calcium concentrations and to allow Mn2 to enter cells to quench indicator fluorescence. However, since 4-bromo A-23187 is nonfluorescent, it is preferred over the blue fluorescent A-23187 for calibrations of UV-excited Ca2 indicators such as Indo-1 and Fura-2 in order to keep background fluorescence to a minimum.


  • White solid soluble in DMSO or EtOH
  • Store at 4°C

Formel: C₂₉H₃₆BrN₃O₆
Molecular Weight: 602,52 g/mol
CAS-Nummer: 76455-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR