Ionomycin Calciumsalz Ionophor

Lieferant: Biotium
Achtung

59007
59007.EA 303 EUR
59007.
Ionomycin Calciumsalz Ionophor
Ionomycin calcium salt

Ionomycin is commonly used to modify intracellular calcium concentrations and to calibrate fluorescent calcium indicators. It is also known for its antiproliferative effects.


  • Yellowish oily solid soluble in DMSO or EtOH
  • Store desiccated at ≤ 4°C

Formel: C₄₁H₇₀CaO₉
Lagertemperatur: Kühlschrank
MDL: MFCD00151183
CAS-Nummer: 56092-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR