2-Methylbutan ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
iso-Pentan
59070-1L
59070-1LEA
178.53
EUR
59070-1L
2-Methylbutan ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
2-Methylbutan
Beilstein Number: 1730723
RTECS Number: EK4430000
UNSPSC Code: 12191502
Formel:
CH₃CH₂CH(CH₃)₂ Molecular Weight: 72.15 g/mol Siedepunkt: 28 °C (1.013 hPa) Schmelzpunkt: –160 °C Dichte: 0.62 g/cm³ (20 °C) Flammpunkt: 7.6 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009338 CAS-Nummer: 78-78-4 EINECS: 201-142-8 UN: 1265 ADR: 3 I |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 99.5 % |
Refractive index (n 20/D) | 1.353 - 1.355 |
Water (Karl Fischer) | Max. 0.05 % |
Non-volatile matter | Max. 0.001 % |
Identity (NMR) | complying |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Explosion Limit | 8.3 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...