Thioflavin T fluorescent dye, Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T

596200-500
596200-500EA 126 EUR
596200-500
Thioflavin T fluorescent dye, Calbiochem®, Sigma-Aldrich®
Thioflavin T

A cell-permeable benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid fibrils.


Verpackung: Packaged under inert gas.

Formel: C₁₇H₁₉ClN₂S
Molecular Weight: 318.86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011944
CAS-Nummer: 2390-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR