Magnesiumhydroxid 95,0-100,5, powder_white USP, FCC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: wasserhaltige Magnesia

M.O.C. Category R

5984-25EA 13013.73 EUR
5984-25 5984-12 5984-20
Magnesiumhydroxid 95,0-100,5, powder_white USP, FCC, Macron Fine Chemicals™
Magnesiumhydroxid
Formel: Mg(OH)₂
Molecular Weight: 58.32 g/mol
Dichte: 2.3261…2.3631 g/cm³ (20 °C)
MDL: MFCD00011104
CAS-Nummer: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical 95.0 - 100.5 %
USP - Assay (Mg(OH)₂) (dried basis) 98.0 - 102.0 %
USP - Calcium (Ca) ≤ 1.5 %
USP - Carbonate (CO₃) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
Lead (Pb)(USP) ≤ 1.5 ppm
USP - Loss on Drying at 105°C ≤ 2.0 %
USP - Loss on Ignition 30.0 - 33.0 %
USP - Soluble Salts ≤ 2.0 %
USP - Microbiological E Coli (negative) Passes Test
FCC - Assay (Mg(OH)₂) (dried basis) 95.0 - 100.5 %
FCC - Identification Passes Test
FCC - Loss on Drying at 105°C ≤ 2.0 %
FCC - Loss on Ignition 30.0 - 33.0 %
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Calcium Oxide ≤ 1 %
FCC - Alkalies (as SO₄) Passes Test
Appearance (fine, white powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR