Lithium standard for IC, TraceCERT®, Supelco®
Lieferant: Merck
59878-100ML
59878-100MLEA
60.6
EUR
59878-100ML
Lithium standard for IC, TraceCERT®, Supelco®
Standards
Kationischer Standards
Lithium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034