1,6-Diphenyl-1,3,5-hexatrien Membranfarbstoff, für die Histologie

Lieferant: Biotium
Achtung

Synonyme: DPH, 1,6-Diphenylhexa-1,3,5-trien, (6-Phenyl-1,3,5-hexatrienyl)benzol, (6-Phenyl-1,3,5-hexatrien-1-yl)benzol

60021
60021.EA 102 EUR
60021.
1,6-Diphenyl-1,3,5-hexatrien Membranfarbstoff, für die Histologie
1,6-Diphenyl-1,3,5-hexatrien

1,6-Diphenyl-1,3,5-hexatriene (DPH) is almost non-fluorescent in water, but it exhibits a strong increase in fluorescence after intercalation into membranes and can be used as a probe for viscosity, polarity and lipid order.


  • Colors: Blue
  • λEx/λEm = 350/420 nm
  • Yellow solid soluble in DMF and DMSO
  • Store at -20 °C and protect from light


DPH has been used for the high-content imaging of neutral lipid droplets1 as well as a reporter of inner spore membrane fluidity in bacterial spores.

Formel: (C₆H₅)CHCHCHCHCHCH(C₆H₅)
Molecular Weight: 232,33 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00004793
CAS-Nummer: 1720-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33660a9ec16ec7460a97d95d08d09e99d24e1fbf&vid=33660a9ec16ec7460a97d95d08d09e99d24e1fbf&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR