Biotin-DHPE Membranfarbstoff, für die Histologie

Lieferant: Biotium

Synonyme: Biotin DHPE, N-(Biotinoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt

60022
60022.EA 393 EUR
60022.
Biotin-DHPE Membranfarbstoff, für die Histologie
Biotin-DHPE

Biotin-DHPE (N-(Biotinoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt) can be used to couple avidin or streptavidin to cell membranes or liposomes.

Formel: C₅₃H₁₀₃N₄O₁₀PS
Molecular Weight: 1019,44 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 136235-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75cd3dd4232d001ce51bf857a201a2beb4ad8d6f&vid=75cd3dd4232d001ce51bf857a201a2beb4ad8d6f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR