Fluorescein-DHPE pH-Indikator, membrane stain for histology
Lieferant: Biotium
Fluorescein-DHPE (N-(Fluorescein-5-thiocarbamoyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt) is a useful surface pH indicator (1) and has also been used for measuring lateral diffusion in membranes via fluorescence photobleaching recovery method.
- λEx/λEm (MeOH) = 496/ 519 nm
- Orange solid soluble in chloroform
- Store at -20°C and protect from light
Formel:
C₆₄H₁₀₀N₃O₁₃PS Lagertemperatur: Gefrierschrank |
CAS-Nummer:
87706-98-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...