NBD-PE Membranfarbstoff, für die Histologie

Lieferant: Biotium

Synonyme: N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt

60025
60025.EA 433 EUR
60025.
NBD-PE Membranfarbstoff, für die Histologie
NBD-PE

NBD-PE (N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt) in combination with Rhodamine-DHPE (#60026) or Texas Red®-DHPE (#60027) has been used to study membrane fusion via fluorescence resonance energy transfer (FRET) (1-3). NBD-PE has also been used in photobleaching recovery measurement.


  • λEx/λEm (MeOH) = 463/536 nm
  • Orange solid soluble in chloroform
  • Store at -20°C and protect from light

Formel: C₄₉H₉₀N₅O₁₁P
Molecular Weight: 956,24 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 178119-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7688e6ce39217e41d4d4bf90fbd052016647782f&vid=7688e6ce39217e41d4d4bf90fbd052016647782f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR