TRITC-DHPE Membranfarbstoff, für die Histologie

Lieferant: Biotium

60028
60028.EA 306 EUR
60028.
TRITC-DHPE Membranfarbstoff, für die Histologie
TRITC-DHPE

TRITC-DHPE is a rhodamine-labeled glycerophosphoethanolamine lipid. DHPE conjugates of rhodamine dyes have been used in membrane fusion assays.


  • λEx/λEm(MeOH) = 540/566 nm
  • Dark solid soluble in chloroform
  • Store at -20°C and protect from light


DHPE conjugates of rhodamine dyes have been used as fluorescence energy acceptors in combination with NBDPE as the donor in membrane fusion assays using the principle of fluorescence energy transfer (FRET). These probes have also been used for following membrane trafficking during endocytosis.

Formel: C₆₈H₁₁₀N₅O₁₁PS
Lagertemperatur: Gefrierschrank
CAS-Nummer: 176181-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bfd970b3535c3adefa145fa88e0b0af9c34176a&vid=8bfd970b3535c3adefa145fa88e0b0af9c34176a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR