C6 NBD-Ceramide Membranfarbstoff, für die Histologie
Lieferant: Biotium
Synonyme:
NBD C6-Ceramide, N-[7-(4-nitrobenzo-2-oxa-1,3-diazole)]-6-aminocaproyl-D-erythro-sphingosine, 6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosine, N-C6-NBD-D-erythro-Sphingosine
60030
60030.EA
491
EUR
60030.
C6 NBD-Ceramide Membranfarbstoff, für die Histologie
C6 NBD-Ceramide
NBD C6-ceramide (6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosine) has been used for following sphingolipid metabolism in cells, and as a selective stain for the Golgi apparatus in live and fixed cells.
- λEx/λEm (MeOH) = 466/536 nm
- Dark solid soluble in chloroform or DMSO
- Store at -20°C and protect from light
Formel:
C₃₀H₄₉N₅O₆ Molecular Weight: 575,74 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
94885-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...