NBD C6-Sphingomyelin Membranfarbstoff, für die Histologie
Lieferant: Biotium
Synonyme:
6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosyl phosphocholine, C6 NBD-sphingosylphosphorylcholine, C6 NBD-Sphingomyelin
60031
60031.EA
565
EUR
60031.
NBD C6-Sphingomyelin Membranfarbstoff, für die Histologie
NBD C6-Sphingomyelin
NBD C6-sphingomyelin (6-((N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoyl)sphingosyl phosphocholine) has been used for following sphingolipid metabolism in cells.
- λEx/λEm (MeOH) = 466/536 nm
- Dark solid soluble in chloroform or DMSO
- Store at -20°C and protect from light
Formel:
C₃₅H₆₁N₆O₉P Molecular Weight: 740,87 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
94885-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...