Octadecyl rhodamine B chloride (R 18) Membranfarbstoff, für die Histologie

Lieferant: Biotium
Gefahr

60033
60033.EA 487 EUR
60033.
Octadecyl rhodamine B chloride (R 18) Membranfarbstoff, für die Histologie
Octadecyl rhodamine B chloride

The fluorescence of octadecyl rhodamine B in membranes is quenched at high dye concentration but is released at dilution. This property makes the dye useful for membrane fusion assays.


  • λEx/λEm (MeOH) = 556/578 nm
  • Dark solid soluble in EtOH or DMSO
  • Store at -20°C and protect from light

Formel: C₄₆H₆₇ClN₂O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00467892
CAS-Nummer: 65603-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68dd563879876de1288734de539d167ed6cc19fc&vid=68dd563879876de1288734de539d167ed6cc19fc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR