Kaliumbromat, getrocknet ≥99,8% (Trockenmasse, RT), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

60085-250G 60085-50G
60085-250GEA 92.58 EUR
60085-250G 60085-50G
Kaliumbromat, getrocknet ≥99,8% (Trockenmasse, RT), Puriss. p.a. ACS, Fluka™
Potassium bromate
Formel: KBrO₃
Siedepunkt: 425 °C
Schmelzpunkt: 409 °C
Dichte: 3,270 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011359
CAS-Nummer: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Bestellen

Spezifikation Testergebnisse

Assay (excluding dried substance) 99.8 - 100.5 %
pH (5 %, 25 °C) 5.0 - 9.0
Colour of the solution complying
Turbidity of solution complying
Loss on drying (150°C) Max. 0.1 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Bromide (Br) complying
Sulfate (SO₄) Max. 50 ppm
Total nitrogen (as N) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR