Kaliumbromid ≥99,5% (durch argentometrische Titration), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Achtung

60093-250G 60093-1KG
60093-250GEA 73.31 EUR
60093-250G 60093-1KG
Kaliumbromid ≥99,5% (durch argentometrische Titration), Puriss. p.a. ACS, Fluka™
Kaliumbromid
Formel: KBr
Siedepunkt: 1,383 °C (1.013 hPa)
Schmelzpunkt: 734 °C
Dichte: 2,73 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011358
CAS-Nummer: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Bestellen

Spezifikation Testergebnisse

Assay 99.5 - 101.0 %
pH (5 %, 25 °C) 5.0 - 8.8
Colour of the solution complying
Clearness of the solution complying
Loss on drying (130°C) Max. 0.3 %
Water insoluble matter Max. 50 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 100 ppb
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Bromate (BrO₃) Max. 10 ppm
Chloride (Cl) Max. 1000 ppm
Iodide (I) Max. 10 ppm
Iodate (IO₃) Max. 10 ppm
Sulfate (SO₄) Max. 50 ppm
Total N Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR