Magnesiumoxid 96,0-100,5%, Pulver USP, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
gebrannte Magnesia
Veränderungsmanagement-Kategorie= R
6018-23EA
2721.22
EUR
6018-23
Magnesiumoxid 96,0-100,5%, Pulver USP, Macron Fine Chemicals™
Magnesiumoxid
Formel:
MgO Molecular Weight: 40.3 g/mol Siedepunkt: 3600 Schmelzpunkt: 2800 Dichte: 3.58 g/cm³ (25 °C) |
MDL:
MFCD00011109 CAS-Nummer: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Spezifikation Testergebnisse
GMP Manufactured Product | — |
Meets U.S.P Requirements | — |
CAUTION: For Manufacturing, processing or repackaging | — |
Bulk Pharmaceutical Chemical | — |
USP - Assay (MgO) (ignited basis) | 96.0 - 100.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Acid-Insoluble Substances | ≤ 0.1 % |
USP - Arsenic (FCC) | ≤ 3 mg/kg |
USP - Bulk Density | ≤ 0.2 mg/g |
USP - Calcium Oxide (FCC) | ≤ 1.5 % |
Foreign Matter | ≤ 0.2 mg/g |
USP - Lead (Pb)(FCC) | ≤ 4 mg/kg |
USP - Limit of Calcium (Ca) | ≤ 1.1 % |
USP - Free Alkali and Soluble Salts | ≤ 2.0 % |
USP - Iron (Fe) | ≤ 0.05 % |
USP - Loss on Ignition | ≤ 10.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...