Salpetersäure ≥70%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

6019.9200PEEA 3334.48 EUR
6019.9200PE
Salpetersäure ≥70%, BAKER ANALYZED® ACS, J.T.Baker®
Salpetersäure
Formel: HNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,I

Bestellen

Spezifikation Testergebnisse

Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs.
Assay (acidimetric) 69.0 - 70.0 %
Appearance passes test
Color (APHA) max. 10
Residue after Ignition max. 4 ppm
Specific Gravity at 60°/60°F 1.416 - 1.420
Trace Impurities (in ppm)
Trace Impurities (in ppm) Chloride (Cl) max. 0.1
Trace Impurities (in ppm) Phosphate (PO4) max. 0.2
Trace Impurities (in ppm) Sulfate (SO4) max. 0.5
Trace Impurities (in ppb)
Trace Impurities (in ppb) Arsenic and Antimony (as As) max. 4
Trace Impurities (in ppb) Aluminium (Al) max. 100
Trace Impurities (in ppb) Gold (Au) max. 200
Trace Impurities (in ppb) Boron (B) max. 50
Calcium (Ca) max. 200
Chromium (Cr) max. 100
Copper (Cu) max. 50
Iron (Fe) max. 100
Mercury (Hg) max. 5
Potassium (K) max. 300
Magnesium (Mg) max. 100
Manganese (Mn) max. 100
Nickel (Ni) max. 50
Lead (Pb) max. 100
Tin (Sn) max. 200
Titanium (Ti) max. 200
Zinc (Zn) max. 200
Heavy Metals (as Pb) max. 100


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR