Kaliumdihydrogenphosphat, wasserfrei 99,5-100,5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. Puffersubstanz, Fluka™

Lieferant: Honeywell Chemicals

60220-2.5KG 60220-500G 60220-1KG 60220-5KG
60220-2.5KGEA 240 EUR
60220-2.5KG 60220-500G 60220-1KG 60220-5KG
Kaliumdihydrogenphosphat, wasserfrei 99,5-100,5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. Puffersubstanz, Fluka™
Kaliumdihydrogenphosphat
Formel: KH₂PO₄
Schmelzpunkt: 253 °C
Dichte: 2,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

Assay (alk., calculated to dried substances) 99.5 - 100.5 %
Assay Min. 99.0 %
Loss on drying (130°C) Max. 0.2 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 4.2 - 4.5
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0015 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 5 ppm
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Sodium (Na) Max. 50 ppm
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm
Total N Max. 0.001 %
KMnO₄ reducing matter (as O) complying
Appearance of the solution complying
Appearance (color) complying
Appearance (form) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR