Potassium hydroxide hydrate ≥99,995% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
60371-25G
60371-25GEA
107.39
EUR
60371-25G
Potassium hydroxide hydrate ≥99,995% (Metall-Basis), TraceSELECT™ für die Spurenanalyse, Fluka™
Potassium hydroxide hydrate
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Formel:
H₃KO₂ Molecular Weight: 74.12 g/mol Siedepunkt: 1.320 °C (1.013 hPa) Schmelzpunkt: 143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003553 CAS-Nummer: 26288-25-5 EINECS: 215-181-3 UN: 1813 ADR: 8 II REACH: 01-2119487136-33 |
Spezifikation Testergebnisse
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.05 ppm |
Barium (Ba) | Max. 1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cerium (Ce) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Chromium (Cr) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.01 ppm |
Europium (Eu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.05 ppm |
Mercury (Hg) | Max. 1 ppb |
Iridium (Ir) | Max. 0.5 ppm |
Lanthanum (La) | Max. 0.01 ppm |
Lithium (Li) | Max. 0.5 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.05 ppm |
Sodium (Na) | Max. 5 ppm |
Nickel (Ni) | Max. 0.01 ppm |
Lead (Pb) | Max. 0.01 ppm |
Palladium (Pd) | Max. 0.5 ppm |
Platinum (Pt) | Max. 0.5 ppm |
Rhodium (Rh) | Max. 0.5 ppm |
Ruthenium (Ru) | Max. 0.5 ppm |
Scandium (Sc) | Max. 0.01 ppm |
Samarium (Sm) | Max. 0.01 ppm |
Strontium (Sr) | Max. 0.5 ppm |
Thallium (Tl) | Max. 0.01 ppm |
Vanadium (V) | Max. 0.1 ppm |
Yttrium (Y) | Max. 0.01 ppm |
Ytterbium (Yb) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.05 ppm |
Silicate (as SiO₂) | Max. 5 ppm |
Chloride (Cl) | Max. 1000 ppb |
Phosphate (PO₄) | Max. 50 ppb |
Sulfate (SO₄) | Max. 5000 ppb |
Carbonate (K₂CO₃) | Max. 0.6 % |
Total nitrogen (as N) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...