di-Kaliumoxalat Monohydrat 99,0 RT, Purum p.a. analytical reagent
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Oxalsäure Dikaliumsalz Monohydrat, Oxalsäure Kaliumsalz Monohydrat, Ethandisäure Dikaliumsalz Monohydrat
60425-1KG
60425-250G
60425-1KGEA
135.42
EUR
60425-1KG
60425-250G
di-Kaliumoxalat Monohydrat 99,0 RT, Purum p.a. analytical reagent
di-Kaliumoxalat Monohydrat
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Formel:
(COOK)₂·H₂O Molecular Weight: 184.23 g/mol Siedepunkt: Zers. (beim Erhitzen) Schmelzpunkt: 356 Dichte: 2.120 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150033 CAS-Nummer: 6487-48-5 EINECS: 209-506-8 Merck Index: 12,07820 |
Spezifikation Testergebnisse
Assay | 99.0 - 101.0 % |
pH (5 %, 20°C) | 7.0 - 8.5 |
Calcium (Ca) | Max. 50 ppm |
Cadmium (Cd) | Max. 50 ppm |
Cobalt (Co) | Max. 50 ppm |
Copper (Cu) | Max. 50 ppm |
Iron (Fe) | Max. 50 ppm |
Sodium (Na) | Max. 5000 ppm |
Nickel (Ni) | Max. 50 ppm |
Lead (Pb) | Max. 50 ppm |
Zinc (Zn) | Max. 50 ppm |
Chloride (Cl) | Max. 50 ppm |
Sulfate (SO₄) | Max. 200 ppm |
Colour of the solution | complying |
Turbidity of solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...