2,2,4-Trimethylpentan 99,5, ChromAR® LC, UV, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Isooktan, Isobutyltrimethylmethan

6043-25EA 179.8 EUR
6043-25
2,2,4-Trimethylpentan 99,5, ChromAR® LC, UV, Macron Fine Chemicals™
Isooctan
Formel: (CH₃)₂CHCH₂C(CH₃)₃
Molecular Weight: 114.23 g/mol
Siedepunkt: 99.2
Schmelzpunkt: –107 °C
Dichte: 0.692 g/cm³ (20 °C)
Flammpunkt: –12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008943
CAS-Nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0003 %
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.02 %
ACS - Water-Soluble Titrable Acid (meq/g) ≤0.0003
Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 - 400 nm ≤0.01
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR