Human thrombin (from plasma)
Lieferant: Merck Millipore Calbiochem
605195-100 605195-5000
605195-100EA 142 EUR
605195-100 605195-5000
Human thrombin (from plasma)
Enzyme
Designed for use in thrombin clotting time tests. Key enzyme that activates factor XIII and converts fibrinogen into fibrin.