Schwefelsäure 95-97%, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

6057.1000EA 55.2 EUR
6057.1000 6057.2500 6057.9025 6057.2500PE 6057.9200
Schwefelsäure 95-97%, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Schwefelsäure
Formel: H₂SO₄
Molecular Weight: 98.08 g/mol
Siedepunkt: ∼335 °C (1013 hPa)
Schmelzpunkt: 10,38
Dichte: 1.84 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Assay 95 - 97 %
Ammonium (NH4) max. 1 ppm
Color (APHA) max. 10
Heavy Metals (as Pb) max. 5 ppm
Nitrate (NO3) max. 1 ppm
Residue after Ignition max. 5 ppm
Substances Reducing KMnO4(as SO2) max. 5 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.05
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Chloride (Cl) max. 0.5
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.01
Iron (Fe) max. 0.1
Germanium (Ge) max. 0.05
Potassium (K) max. 0.1
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.05
Zirconium (Zr) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR