Schwefelsäure 95-97%, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
6057.1000EA
55.2
EUR
6057.1000
6057.2500
6057.9025
6057.2500PE
6057.9200
Schwefelsäure 95-97%, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Schwefelsäure
Formel:
H₂SO₄ Molecular Weight: 98.08 g/mol Siedepunkt: ∼335 °C (1013 hPa) Schmelzpunkt: 10,38 Dichte: 1.84 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064589 CAS-Nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Spezifikation Testergebnisse
Assay | 95 - 97 % |
Ammonium (NH4) | max. 1 ppm |
Color (APHA) | max. 10 |
Heavy Metals (as Pb) | max. 5 ppm |
Nitrate (NO3) | max. 1 ppm |
Residue after Ignition | max. 5 ppm |
Substances Reducing KMnO4(as SO2) | max. 5 ppm |
Trace Impurities (in ppm) | |
Silver (Ag) | max. 0.01 |
Aluminium (Al) | max. 0.05 |
Arsenic (As) | max. 0.05 |
Barium (Ba) | max. 0.02 |
Beryllium (Be) | max. 0.01 |
Calcium (Ca) | max. 0.5 |
Cadmium (Cd) | max. 0.02 |
Chloride (Cl) | max. 0.5 |
Cobalt (Co) | max. 0.01 |
Chromium (Cr) | max. 0.1 |
Copper (Cu) | max. 0.01 |
Iron (Fe) | max. 0.1 |
Germanium (Ge) | max. 0.05 |
Potassium (K) | max. 0.1 |
Lithium (Li) | max. 0.01 |
Magnesium (Mg) | max. 0.1 |
Manganese (Mn) | max. 0.01 |
Molybdenum (Mo) | max. 0.02 |
Sodium (Na) | max. 0.5 |
Nickel (Ni) | max. 0.05 |
Lead (Pb) | max. 0.05 |
Strontium (Sr) | max. 0.01 |
Titanium (Ti) | max. 0.1 |
Thallium (Tl) | max. 0.05 |
Vanadium (V) | max. 0.01 |
Zinc (Zn) | max. 0.05 |
Zirconium (Zr) | max. 0.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...