Kieselgel, Porengröße: 60 Å Ca, ~0.1% 230-400 mesh, hochrein, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
60752-1KG
60752-5KG
60752-1KGEA
210
EUR
60752-1KG
60752-5KG
Kieselgel, Porengröße: 60 Å Ca, ~0.1% 230-400 mesh, hochrein, Supelco®
Silica gel
Formel:
SiO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 112926-00-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...