Desk timer/stopwatch
Lieferant: EA COMBS
5010
609-0002EA 176 EUR
609-0002
Desk timer/stopwatch
Timer Timer, digital
Bench top clock/stopwatch with count up/down timer.
  • Four count up timing channels: seconds, minutes, hours; decimal seconds; ¹/₁₀₀ seconds; decimal minutes
  • Countdown timer from 24 hours with memory and alarm at end of period
  • Clock with calendar and daily alarm
  • Accuracy within one second over a 24 hour period
Lieferumfang: Supplied with R14 battery.