VWR® Traceable® Calibrated Timer
Lieferant: VWR Collection
609-0195EA 65.5 EUR
609-0195
VWR® Traceable® Calibrated Timer
Timer Timer, digital
Personalise with coloured faceplates.
  • Unique memory returns channel to previously programmed setting at touch of a key
  • Four independent timing channels
  • NIST-traceable
  • Counts up during alarming indicating elapsed time since alarming, 12/24 hour clock
  • 1 year warranty
Four independent timing channels feature unique audible alarms for simultaneous timing. Count up or down from 100 hours for longer duration experiments. Memory function returns the display to the programmed time.
Traceable® Products have an individually numbered Traceable® Certificate provided with each unit, that assures accuracy from an ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving users both time and money by not having to calibrate separately.
Guaranteed to have at least one year of calibration remaining on certificate.
Standards: Kalibrierung gemäß ISO/IEC 17025, ANSI/NCSL Z540-1 und 9001, akkreditiert von der A2LA belegt die Rückführbarkeit auf NIST.
Lieferumfang: Lieferung mit Tischständer, magnetischer Rückseite, Batterie und Kalibrierzertifikat.