Mechanical time switch
Lieferant: BACHMANN ELEKTRO
852.101
609-1701EA 17.3 EUR
609-1701
Mechanical time switch
Timer Zeitschaltuhren
Colour: White.
  • Plastic casing
  • Switching interval of 15 minutes
  • Up to 96 switch-on and switch-off times per day (24 hours)