Salpetersäure 65%, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Nitric acid solution

6090.2500PEEA 130 EUR
6090.2500PE
Salpetersäure 65%, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Salpetersäure
Formel: HNO₃
Molecular Weight: 63.01 g/mol
Siedepunkt: 87
Schmelzpunkt: –42 °C
Dichte: 1.4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Bestellen

Spezifikation Testergebnisse

Assay min. 65 %
Dithizone determination passes test
Chloride (Cl) max. 0.5 ppm
Mercury (Hg) max. 5 ppb
Residue after Ignition max. 5 ppm
Sulfate (SO4) max. 1 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.05
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.01
Iron (Fe) max. 0.1
Germanium (Ge) max. 0.05
Potassium (K) max. 0.1
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.05
Zirconium (Zr) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR