Di-4-ANEPPS Membranfarbstoff, für die Histologie
Lieferant: Biotium
Di-4-ANEPPS is a fast-responding membrane potential dye. Changes in the membrane potential of the cell correlate with changes in the fluorescence excitation intensity of the dye.
- λEx/λEm (MeOH) = 496/705 nm*
- Orange red solid soluble in DMF, ethanol or DMSO
- Store at 4°C and protect from light, especially in solution
Di-4-ANEPPS is internalized in cells rapidly, so it is mainly used for short-term experiments. We also offer Di-8-ANEPPS (catalog number 61012), which is more lipophilic and better retained in the outer leaflet of the plasma membrane.
* Note that the spectra of styryl dyes generally undergo a large blue shift in lipids compared to organic solvent. See the spectra for styryl dyes in liposomes under product images.
Formel:
C₂₈H₃₆N₂O₃S Lagertemperatur: Kühlschrank |
MDL:
MFCD03453609 CAS-Nummer: 90134-00-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...