RH 414 Membranfarbstoff, für die Histologie
Lieferant: Biotium
Synonyme:
N-(3-Triethylammoniumpropyl)-4-(4-(4-(diethylamino)phenyl)butadienyl)pyridinium dibromide
61016
61016.EA
480
EUR
61016.
RH 414 Membranfarbstoff, für die Histologie
RH 414
The styryl dye RH414 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons.
- λEx/λEm (MeOH) = 532/716 nm
- Orange red soluble in H2O or DMSO
- Store at -20°C and protect from light
Excitation/emission data is for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission.
Formel:
C₂₈H₄₃Br₂N₃ Molecular Weight: 581,476 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
161433-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...