RH421 Membranfarbstoff, für die Histologie

Lieferant: Biotium

Synonyme: 4-{4-[4-(Dipentylamino)phenyl]-1,3-butadienyl}-1-(4-sulphobutyl)pyridinium hydroxide, N-(4-sulphobutyl)-4-(4-(4-(dipentylamino)phenyl)butadienyl) pyridinium, inner salt

61017
61017.EA 480 EUR
61017.
RH421 Membranfarbstoff, für die Histologie
RH421

RH421 is a fast responding membrane potential dye, exhibiting >20% fluorescence change per 100 mV on neuroblastoma cells.


  • λExλEm= 515/704 nm (MeOH)
  • Red solid soluble in DMSO, EtOH or DMF
  • Store at -20°C and protect from light, especially in solution

Formel: C₂₉H₄₂N₂O₃S
Molecular Weight: 498,72 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 107610-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de82cf79896cf94b9fcfc4424097f444510626c5&vid=de82cf79896cf94b9fcfc4424097f444510626c5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR