p-Kresol ≥99,0% (durch GC), Puriss. p.a., Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
61030-25G
61030-25GEA
61.62
EUR
61030-25G
p-Kresol ≥99,0% (durch GC), Puriss. p.a., Riedel-de Haën™
p-Kresol
Beilstein Number: 1305151
RTECS Number: GO6475000
UNSPSC Code: 12190000
Formel:
C₇H₈O Siedepunkt: 202 °C (1,013 hPa) Schmelzpunkt: 35,5 °C (1.013 hPa) Dichte: 1,03 g/cm³ (20 °C) Flammpunkt: 86,0 °C (1.013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002376 CAS-Nummer: 106-44-5 EINECS: 203-398-6 UN: 3455 ADR: 6.1,II Merck Index: 13,02604 |
Spezifikation Testergebnisse
assay (alkalimetric) | Min. 99 % |
identity (IR) | complying |
appearance | complying |
Cadmium (Cd) | Max. 5 ppm |
APHA | Max. 25 |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 5 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...