Calibration weight sets, class M1
Lieferant: ZWIEBEL
P8134612
611-0965EA 0 EUR
611-0965
Calibration weight sets, class M1
Gewichte Kalibriergewichte
These masses are intended for use calibrating of weights of a lower accuracy class and for use with weighing instruments of medium accuracy class III and and ordinary accuracy class IIII.
Standards: Conform to the requirements of OIML recommendation R111.