Halbmikrowaagen, Explorer®-Serie
Lieferant: OHAUS
30139517 30139515 30139516 30139518 30139519 30218998 30218999 30219000 30219001 30219002
611-3321EA 8220 EUR
611-3321 611-3319 611-3320 611-3322 611-3323 611-3312 611-3313 611-3314 611-3315 611-3316
Halbmikrowaagen, Explorer®-Serie
Waagen
These semi-microbalances have been designed and built for sophisticated professional weighing applications where the user requires extreme accuracy.
  • 'High Speed Single Module' weighing cell delivers ultra-fast stabilisation times typically ≤3 seconds for 0,1 mg and ≤8 seconds for 0,01 mg
  • Intuitive software with an icon-based colour touch screen display
  • AutoCal™ automatic internal calibration which uses two internal weights to perform a linear calibration
  • Save data to USB
  • Four touchless sensors allow the operator to perform common functions and commands with a swipe of the hand
  • Choice of anti-static glass draft shield with flip-top door, or Auto-Door - automatic draft shield doors for access to chamber without the need to touch the doors, minimising unwanted vibration
  • Built-in ioniser with Auto-Door models
  • LIMS compatibility
Die Nutzer-Manager-Funktion umfasst Profile für einen Administrator sowie fünf weitere Benutzer mit der Anzeige einer Probe unter Mindesteinwaage und Echtzeituhr für die GLP/GMP-Datenerfassung.
Bestellinformation: A selection of OIML EC approved balances denoted M (e.g., EX125DM) are also available. For the full range of accessories for the Explorer® semi-micro balance please contact your local Avantor sales office.