Feuchtebestimmer, professionell, MB 90 und MB 120
Lieferant: OHAUS
30246943 30246944 30241169 30303303 30303304
611-3579EA 3350 EUR
611-3579 611-3580 611-3571 611-3746 611-3747
Feuchtebestimmer, professionell, MB 90 und MB 120
Analysatoren Feuchte-Analysatoren
Moisture analysers simplify complex moisture analysis applications. Designed to increase precision and efficiency in the laboratory, they feature a precisely controlled halogen heating system which gives fast, accurate results.
  • Colour touch screen display with intuitive icon-based navigation
  • Easy, tool-free cleaning for convenient maintenance
  • Quickly connects to a PC or flash drive
  • Precisely controlled, uniform, fast halogen heating technology provides repeatable results
  • Define drying parameters and store up to 100 methods with MB 120
MB 90 is suitable for routine moisture analysis applications, the units, halogen heating begins to dry samples in seconds and performs up to 40% faster than traditional infrared methods.
MB 120 is more adapted to professional moisture analysis, with advanced features, including the new Temperature Guide which analyses a sample to determine the optimum drying temperature, and a Methods Library that can store up to 100 methods and 1000 total results. In addition, there are four selectable drying profiles for easy one-step drying and seven shut-off criteria to automatically end the test (once moisture loss is no longer detected). This high performance moisture analyser can test a variety of sample types, providing precise, repeatable results helping to streamline the daily workflow, and offers three levels of user management to meet individual user requirements and ensure data safety.
Standards: CE, UL listed, CSA and FCC approved.