Kjeldahl analysis weighing boats, Grade 609, Whatman™
Lieferant: Whatman products (Cytiva)
10313032
611-3990EA 136 EUR
611-3990
Kjeldahl analysis weighing boats, Grade 609, Whatman™
Schiffchen Wägeschiffchen
Cytiva's Whatman parchment weighing boats are designed so users can collect, transfer and drop both sample and weighing boat into the acid solution during a Kjeldahl analysis without influencing analytical results.
  • A fast and reliable way to safely transfer samples for Kjeldahl analysis
  • Excellent for both weighing and quantitative sample transfer
  • Made from low nitrogen parchment paper
  • No glue or chemical additives
  • Weighing boats dissolve residue-free in the digestion solution without influencing analytical results
Cytiva's business offers weighing papers for Kjeldahl analysis.
Weight 80 g/m²